Ethyl isovalerate, 98%, Thermo Scientific Chemicals
Ethyl isovalerate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl isovalerate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 mL
100 mL
500 mL
Catalog number A10292.0F
also known as A10292-0F
Price (USD)/ Each
213.00
-
Add to cart
Quantity:
2500 mL
Request bulk or custom format
Price (USD)/ Each
213.00
Add to cart
Ethyl isovalerate, 98%, Thermo Scientific Chemicals
Catalog numberA10292.0F
Price (USD)/ Each
213.00
-
Add to cart
Chemical Identifiers
CAS108-64-5
IUPAC Nameethyl 3-methylbutanoate
Molecular FormulaC7H14O2
InChI KeyPPXUHEORWJQRHJ-UHFFFAOYSA-N
SMILESCCOC(=O)CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3955-1.3995 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only