Thermo Scientific Chemicals

1,4-Dinitrobenzene, 98+%, Thermo Scientific Chemicals

Catalog number: A10293.14
25 g, Each
Thermo Scientific Chemicals

1,4-Dinitrobenzene, 98+%, Thermo Scientific Chemicals

Catalog number: A10293.14
25 g, Each
Quantity
Catalog number: A10293.14
also known as A10293-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-25-4
IUPAC Name
1,4-dinitrobenzene
Molecular Formula
C6H4N2O4
InChI Key
FYFDQJRXFWGIBS-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(C=C1)[N+]([O-])=O
Form
Crystalline powder or crystals or powder
Assay (GC)
≥98%
Identification (FTIR)
Conforms
Appearance (Color)
Pale yellow or orange to brown

Description

1,4-Dinitrobenzene is used in a study to evaluate the ionization mechanism and solvent effect by novel atmospheric pressure photoionization mass spectrometry in negative ion mode for analysis of some compounds. 1,4-Dinitrobenzene can be used in synthesis of dyes and dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dinitrobenzene is used in a study to evaluate the ionization mechanism and solvent effect by novel atmospheric pressure photoionization mass spectrometry in negative ion mode for analysis of some compounds. 1,4-Dinitrobenzene can be used in synthesis of dyes and dye intermediates.

Solubility
Soluble in water. (0.8 g/L) at 20°C.

Notes
Incompatible with bases, alkali metals, oxidizing agents and nitric acid.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text