Thermo Scientific Chemicals

Propargyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A10295.AP
500 mL, Each
Thermo Scientific Chemicals

Propargyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A10295.AP
500 mL, Each
Quantity
Catalog number: A10295.AP
also known as A10295-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-19-7
IUPAC Name
prop-2-yn-1-ol
Molecular Formula
C3H4O
InChI Key
TVDSBUOJIPERQY-UHFFFAOYSA-N
SMILES
OCC#C
Assay (GC)
≥98.5%
Refractive Index
1.4315-1.4345 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Water Content (Karl Fischer Titration)
≤0.5%
Form
Liquid

Description

Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.

Solubility
Fully miscible in water.

Notes
Store at 4°C. Incompatible with oxidizing agent, strong acids, strong bases, amines, phosphorous pentoxide, isocyanate vapor & heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text