Thermo Scientific Chemicals

Benzoin methyl ether, 97%, Thermo Scientific Chemicals

Catalog number: A10296.36
500 g, Each
Thermo Scientific Chemicals

Benzoin methyl ether, 97%, Thermo Scientific Chemicals

Catalog number: A10296.36
500 g, Each
Quantity
Catalog number: A10296.36
also known as A10296-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3524-62-7
IUPAC Name
2-methoxy-1,2-diphenylethan-1-one
Molecular Formula
C15H14O2
InChI Key
BQZJOQXSCSZQPS-UHFFFAOYNA-N
SMILES
COC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream
Melting Point (clear melt)
41.0-51.0°C
Form
Crystals or powder or crystalline powder or lumps
Assay (GC)
≥96.0%

Description

Benzoin methyl ether is used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing , paper varnish and other surface coatings. It is used for anticratering in powder coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoin methyl ether is used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing , paper varnish and other surface coatings. It is used for anticratering in powder coatings.

Solubility
Soluble in methanol.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text