Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A10300.06
5 g, Each
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A10300.06
5 g, Each
Quantity
Catalog number: A10300.06
also known as A10300-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39234-86-1
IUPAC Name
3,5-bis(trifluoromethyl)benzene-1-sulfonyl chloride
Molecular Formula
C8H3ClF6O2S
InChI Key
BTRCVKADYDVSLI-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC(=CC(=C1)C(F)(F)F)S(Cl)(=O)=O
Appearance (Color)
White to pale cream to cream to yellow
Form
Crystals or powder or crystalline powder or fused solid or clear liquid as melt
Assay (GC)
≥96.0%
Refractive Index
1.4435-1.4485 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text