Thermo Scientific Chemicals

Valeronitrile, 99%, Thermo Scientific Chemicals

Catalog number: A10306.AP
500 mL, Each
Thermo Scientific Chemicals

Valeronitrile, 99%, Thermo Scientific Chemicals

Catalog number: A10306.AP
500 mL, Each
Quantity
Catalog number: A10306.AP
also known as A10306-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-59-8
IUPAC Name
pentanenitrile
Molecular Formula
C5H9N
InChI Key
RFFFKMOABOFIDF-UHFFFAOYSA-N
SMILES
CCCCC#N
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.3960-1.3990 @ 20?C

Description

Valeronitrile is used in the preparation of valeric acid. It is also used to enhance the nitrilase activity in many strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valeronitrile is used in the preparation of valeric acid. It is also used to enhance the nitrilase activity in many strains.

Solubility
Soluble in water at 20°C 10g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents, acids, bases and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text