It is used for research or analysis use only, as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used for research or analysis use only, as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, acids.
RUO – Research Use Only
J. W. Collette, D. M. Gale and. G. Belletete. Effect of cyclobutane carbonitrile rings on the glass transition temperature of vinyl copolymers. Journal of Polymer Science.1980, 18 1071-1078.