Thermo Scientific Chemicals

Cyclobutanecarbonitrile, 97%, Thermo Scientific Chemicals

Catalog number: A10309.14
25 g, Each
Thermo Scientific Chemicals

Cyclobutanecarbonitrile, 97%, Thermo Scientific Chemicals

Catalog number: A10309.14
25 g, Each
Quantity
Catalog number: A10309.14
also known as A10309-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4426-11-3
IUPAC Name
cyclobutanecarbonitrile
Molecular Formula
C5H7N
InChI Key
GFBLFDSCAKHHGX-UHFFFAOYSA-N
SMILES
N#CC1CCC1
Appearance (Color)
Clear colorless
Refractive Index
1.4300-1.4350 @ 20?C
Form
Liquid
Assay (GC)
≥96.0%

Description

It is used for research or analysis use only, as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for research or analysis use only, as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text