Thermo Scientific Chemicals

N-Chlorosuccinimide, 98%, Thermo Scientific Chemicals

Catalog number: A10310.0B
1000 g, Each
Thermo Scientific Chemicals

N-Chlorosuccinimide, 98%, Thermo Scientific Chemicals

Catalog number: A10310.0B
1000 g, Each
Quantity
Catalog number: A10310.0B
also known as A10310-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
128-09-6
IUPAC Name
1-chloropyrrolidine-2,5-dione
Molecular Formula
C4H4ClNO2
InChI Key
JRNVZBWKYDBUCA-UHFFFAOYSA-N
SMILES
ClN1C(=O)CCC1=O
Assay from Supplier's CofA
≥97.5% (Iodometric titration, US-sourced material)
Melting Point (clear melt)
144-153?C
Solution Test
Colorless to pale yellow, clear or some fine fibres may be present: [5% in Acetone]
Assay (Iodometric Titration)
≥97.5 to ≤102.5% (UK-sourced material)
Comment
Material sourced in UK and US

Description

Regioselective chlorination and oxidizing reagentN-Chlorosuccinimide is used as a chlorinating agent, mild oxidant and swimming pool disinfectant. It is used in the microdetermination of hydrazine salts by titrimetry as well as in the detection of arsenic in the +3 oxidative state. It is also employed in the production of rubber additives. Further, it is used as pharmaceutical intermediate and antibiotic drugs intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Regioselective chlorination and oxidizing reagentN-Chlorosuccinimide is used as a chlorinating agent, mild oxidant and swimming pool disinfectant. It is used in the microdetermination of hydrazine salts by titrimetry as well as in the detection of arsenic in the +3 oxidative state. It is also employed in the production of rubber additives. Further, it is used as pharmaceutical intermediate and antibiotic drugs intermediate.

Solubility
Soluble in water, alcohol, benzene, acetone and acetic acid. Slightly soluble in ether, chloroform, carbon tetrachloride and petroleum ether.

Notes
Light sensitive and moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, amines, ammonia, iron and iron salts.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text