Thermo Scientific Chemicals

N-Hydroxysuccinimide, 98+%, Thermo Scientific Chemicals

Catalog number: A10312.36
500 g, Each
Thermo Scientific Chemicals

N-Hydroxysuccinimide, 98+%, Thermo Scientific Chemicals

Catalog number: A10312.36
500 g, Each
Quantity
Catalog number: A10312.36
also known as A10312-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6066-82-6
IUPAC Name
1-hydroxypyrrolidine-2,5-dione
Molecular Formula
C4H5NO3
InChI Key
NQTADLQHYWFPDB-UHFFFAOYSA-N
SMILES
ON1C(=O)CCC1=O
Assay from Supplier's CofA
≥98.0% (U.S. specification)
Water Content (Karl Fischer Titration)
≤1.0%
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0% (non-U.S. specification)
Identification (FTIR)
Conforms (non-U.S. specification)
Appearance (Color)
White

Description

N-Hydroxysuccinimide is used for improved amidations in the carbodiimide method. It is also used to activate a carboxyl group and reacts with amine to form amide. It is involved in the preparation of N-hydroxymaleimide-styrene copolymer. Further, it finds use in analytical chemistry. As an additive, it is used in the carbodiimide method for improved amidations and peptide couplings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Hydroxysuccinimide is used for improved amidations in the carbodiimide method. It is also used to activate a carboxyl group and reacts with amine to form amide. It is involved in the preparation of N-hydroxymaleimide-styrene copolymer. Further, it finds use in analytical chemistry. As an additive, it is used in the carbodiimide method for improved amidations and peptide couplings.

Solubility
Soluble in water, dimethyl formamide, alcohols and ethyl acetate. Insoluble in cold ether.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides and strong bases. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text