cis-3-Hexen-1-ol, 98%, Thermo Scientific Chemicals
cis-3-Hexen-1-ol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-3-Hexen-1-ol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number A10313.18
also known as A10313-18
Price (USD)/ Each
139.65
Online exclusive
155.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
139.65
Online exclusive
155.00 
Save 15.35 (10%)
Add to cart
cis-3-Hexen-1-ol, 98%, Thermo Scientific Chemicals
Catalog numberA10313.18
Price (USD)/ Each
139.65
Online exclusive
155.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS928-96-1
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4385-1.4425 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only