3-Aminophthalic acid hydrochloride dihydrate, 96%, Thermo Scientific Chemicals
3-Aminophthalic acid hydrochloride dihydrate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminophthalic acid hydrochloride dihydrate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number A10319.14
also known as A10319-14
Price (USD)/ Each
456.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
456.00
Add to cart
3-Aminophthalic acid hydrochloride dihydrate, 96%, Thermo Scientific Chemicals
Catalog numberA10319.14
Price (USD)/ Each
456.00
-
Add to cart
Chemical Identifiers
CAS1852533-96-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale grey to pale pink
FormPowder
Assay (Titration ex Chloride)≥95.0 to ≤105.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only