Thermo Scientific Chemicals

Triethylsilane, 98+%, Thermo Scientific Chemicals

Catalog number: A10320.09
10 g, Each
Thermo Scientific Chemicals

Triethylsilane, 98+%, Thermo Scientific Chemicals

Catalog number: A10320.09
10 g, Each
Quantity
Catalog number: A10320.09
also known as A10320-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
617-86-7
IUPAC Name
triethylsilyl
Molecular Formula
C6H15Si
InChI Key
QXTIBZLKQPJVII-UHFFFAOYSA-N
SMILES
CC[Si](CC)CC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.4100-1.4140 @ 20?C

Description

Triethylsilane is used for the hydrosilation of olefins to prepare alkyl silanes. It acts as a reducing agent in the reduction of 2-chromanols, since it has an active hydride. It acts as a catalyst for redox initiated cationic polymerization, regioselective reductive coupling of enones and allenes, and Beckmann rearrangement of cyclododecanone oxime. It is associated with trifluoroacetic acid and involved in the selective reduction of alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylsilane is used for the hydrosilation of olefins to prepare alkyl silanes. It acts as a reducing agent in the reduction of 2-chromanols, since it has an active hydride. It acts as a catalyst for redox initiated cationic polymerization, regioselective reductive coupling of enones and allenes, and Beckmann rearrangement of cyclododecanone oxime. It is associated with trifluoroacetic acid and involved in the selective reduction of alkenes.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, water and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text