Thermo Scientific Chemicals

Bisphenol A, 97+%, Thermo Scientific Chemicals

Catalog number: A10324.0B
1000 g, Each
Thermo Scientific Chemicals

Bisphenol A, 97+%, Thermo Scientific Chemicals

Catalog number: A10324.0B
1000 g, Each
Quantity
Catalog number: A10324.0B
also known as A10324-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80-05-7
IUPAC Name
4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
Molecular Formula
C15H16O2
InChI Key
IISBACLAFKSPIT-UHFFFAOYSA-N
SMILES
CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Form
Powder or fused solid or granules or prills
Melting Point (clear melt)
152.0-162.0?C
Appearance (Color)
White
Assay (GC)
≥97.0%
Identification (FTIR)
Conforms

Description

Bisphenol A is an important monomer used in the production of polycarbonate plastics which is utilized to make various consumer goods like water bottles, sports equipment, medical devices, eyeglass lenses, house hold electronics, compact discs (CDs) and digital versatile discs (DVDs). Bisphenol A containing epoxy resins are used to make lining in water pipes. It is used as a precursor to prepare flame retardant tetrabromobispheonl A, polysulfones and polyether ketones, which are antioxidants in some plasticizers. It acts as a polymerization inhibitor in poly vinyl chloride (PVC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bisphenol A is an important monomer used in the production of polycarbonate plastics which is utilized to make various consumer goods like water bottles, sports equipment, medical devices, eyeglass lenses, house hold electronics, compact discs (CDs) and digital versatile discs (DVDs). Bisphenol A containing epoxy resins are used to make lining in water pipes. It is used as a precursor to prepare flame retardant tetrabromobispheonl A, polysulfones and polyether ketones, which are antioxidants in some plasticizers. It acts as a polymerization inhibitor in poly vinyl chloride (PVC).

Solubility
Soluble in aqueous alkaline solution, acetic acid, alcohol and acetone. Slightly soluble in carbon tetrachloride. It has poor solubility in water.

Notes
Incompatible with acid anhydrides, acid chlorides, strong bases and strong oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text