Thermo Scientific Chemicals

Methyl chloroacetate, 99+%, Thermo Scientific Chemicals

Catalog number: A10325.30
250 g, Each
Thermo Scientific Chemicals

Methyl chloroacetate, 99+%, Thermo Scientific Chemicals

Catalog number: A10325.30
250 g, Each
Quantity
Catalog number: A10325.30
also known as A10325-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-34-4
IUPAC Name
methyl 2-chloroacetate
Molecular Formula
C3H5ClO2
InChI Key
QABLOFMHHSOFRJ-UHFFFAOYSA-N
SMILES
COC(=O)CCl
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥99.0%
Refractive Index
1.4205-1.4225 @ 20?C

Description

Methyl chloroacetate is used as a solvent and chemical intermediate. It acts as a precursor in the preparation of (carboxymethyl) trimethylammonium chloride esters. Further, it is used in the preparation of octakis-(carbethoxymethoxy)calix[8]arene. It is employed as an extraction solvent during the separation of neutral compounds. In addition to this, it is used in the synthesis of dimethyl carbonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl chloroacetate is used as a solvent and chemical intermediate. It acts as a precursor in the preparation of (carboxymethyl) trimethylammonium chloride esters. Further, it is used in the preparation of octakis-(carbethoxymethoxy)calix[8]arene. It is employed as an extraction solvent during the separation of neutral compounds. In addition to this, it is used in the synthesis of dimethyl carbonate.

Solubility
Miscible with organic solvents, alcohol, ether, acetone and benzene. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text