Thermo Scientific Chemicals

Magnesium nitrate hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A10329.0I
5000 g, Each
Thermo Scientific Chemicals

Magnesium nitrate hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A10329.0I
5000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10329.0I
also known as A10329-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13446-18-9
IUPAC Name
magnesium(2+) hexahydrate dinitrate
Molecular Formula
H12MgN2O12
InChI Key
MFUVDXOKPBAHMC-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Form
Crystals or powder or crystalline powder or compacted powder
Complexometric Titration
≥97.5 to ≤102.5%
Appearance (Color)
White

Description

It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.

Solubility
Very soluble in water, alcohol and liquid ammonia.

Notes
Heating of magnesium nitrate hexahydrate does not lead to dehydration but decomposition to produce magnesium oxide, nitrogen oxide and oxygen.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text