It is used in the identification of sulfation sites of metabolites and prediction of the compounds’ biological effects and in chemical investigation of Wedelia calendulacea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the identification of sulfation sites of metabolites and prediction of the compounds’ biological effects and in chemical investigation of Wedelia calendulacea.
Solubility
Solubility in methanol is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
- Lin Yi, et al. Identification of sulfation sites of metabolites and prediction of the compounds’ biological effects.Analytical and Bioanalytical Chemistry.,2006,386(3), 666-674.
- T. R. Govindachari, et al. Chemical investigation of Wedelia calendulacea. Part II. The position of the methoxyl group in wedelolactone.J. Chem. Soc.,1957,545-547.