3-Chlorobenzylideneacetone, 98%, Thermo Scientific Chemicals
3-Chlorobenzylideneacetone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chlorobenzylideneacetone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A10332.06
also known as A10332-06
Price (USD)/ Each
92.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
92.40
Add to cart
3-Chlorobenzylideneacetone, 98%, Thermo Scientific Chemicals
Catalog numberA10332.06
Price (USD)/ Each
92.40
-
Add to cart
Chemical Identifiers
CAS20766-36-3
IUPAC Name(3E)-4-(3-chlorophenyl)but-3-en-2-one
Molecular FormulaC10H9ClO
InChI KeyVWEPXSRBBXPYSM-AATRIKPKSA-N
SMILESCC(=O)C=CC1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5950-1.6030 @ 20°C
Appearance (Color)Clear yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only