Thermo Scientific Chemicals

n-Pentadecane, 99%, Thermo Scientific Chemicals

Catalog number: A10336.14
25 g, Each
Thermo Scientific Chemicals

n-Pentadecane, 99%, Thermo Scientific Chemicals

Catalog number: A10336.14
25 g, Each
Quantity
Catalog number: A10336.14
also known as A10336-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
629-62-9
IUPAC Name
pentadecane
Molecular Formula
C15H32
InChI Key
YCOZIPAWZNQLMR-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5% (non-U.S. sourced material)
Assay from Supplier's CofA
≥98.5% (GC, U.S. sourced material)
Comment
Material sourced in the U.S. and in other countries

Description

n-pentadecane is used as a reference substance for gas chromatography. It is also used as standard materials for the determination of combustion quality of diesel. It is involved in the organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-pentadecane is used as a reference substance for gas chromatography. It is also used as standard materials for the determination of combustion quality of diesel. It is involved in the organic synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text