Thermo Scientific Chemicals

N,N-Dimethyl-p-phenylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A10343.36
500 g, Each
Thermo Scientific Chemicals

N,N-Dimethyl-p-phenylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A10343.36
500 g, Each
Quantity
Catalog number: A10343.36
also known as A10343-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
536-46-9
IUPAC Name
dihydrogen N1,N4-dimethylbenzene-1,4-diamine dichloride
Molecular Formula
C8H14Cl2N2
InChI Key
PXJHVKRLFWZUNV-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].CNC1=CC=C(NC)C=C1
Comment
Material may darken on storage
Comment
May contain a small quantity of discolored material from the manufacturing process
Appearance (Color)
White to brown or dark purple to purple to pink to dark gray to gray
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

It is a stain used in microscopy. N,N-Dimethyl-p-phenylenediamine dihydrochloride is also suitable for use in the peroxidase test. It is a compound that is normally used to measure the antioxidant potential.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a stain used in microscopy. N,N-Dimethyl-p-phenylenediamine dihydrochloride is also suitable for use in the peroxidase test. It is a compound that is normally used to measure the antioxidant potential.

Solubility
Soluble in water

Notes
Store away from oxidizing agents, humidity and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from light. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text