Benzyltri-n-butylammonium chloride, 98%, Thermo Scientific Chemicals
Benzyltri-n-butylammonium chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyltri-n-butylammonium chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number A10345.36
also known as A10345-36
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
Add to cart
Benzyltri-n-butylammonium chloride, 98%, Thermo Scientific Chemicals
Catalog numberA10345.36
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS23616-79-7
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)≤1%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only