Thermo Scientific Chemicals

Benzaldehyde, 99+%, Thermo Scientific Chemicals

Catalog number: A10348.0I
5000 g, Each
Thermo Scientific Chemicals

Benzaldehyde, 99+%, Thermo Scientific Chemicals

Catalog number: A10348.0I
5000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10348.0I
also known as A10348-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-52-7
IUPAC Name
benzaldehyde
Molecular Formula
C7H6O
InChI Key
HUMNYLRZRPPJDN-UHFFFAOYSA-N
SMILES
O=CC1=CC=CC=C1
Comment
Purchased in U.K. and U.S.
Free acid (titration)
<0.5% (as Benzoic acid) (U.K. specification)
Identification (FTIR)
Conforms (U.K. specification)
Refractive Index
1.5440-1.5460 @ 20°C (U.K. specification)
Appearance (Color)
Clear colorless to pale yellow

Description

Benzaldehyde is commonly employed as a precursor to prepare various organic compounds, which are used as pharmaceuticals, dyes, perfumes, flavorings, photographic chemicals and plastic additives. It is used as solvent for oils and resins. Benzaldehyde is further used as a building block for a range of amino acids like phenylglycine, which is used as a side chain in various antibiotics. It is involved as a brightener in zinc electroplating processes, and as a food and fragrance additive. It is active intermediate for the manufacture of benzoic acid, odorants and flavoring chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text