Thermo Scientific Chemicals

N-Boc-hydroxylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A10349.06
5 g, Each
Thermo Scientific Chemicals

N-Boc-hydroxylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A10349.06
5 g, Each
Quantity
Catalog number: A10349.06
also known as A10349-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36016-38-3
IUPAC Name
tert-butyl N-hydroxycarbamate
Molecular Formula
C5H11NO3
InChI Key
DRDVJQOGFWAVLH-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NO
Assay (GC)
≥98.0%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
52.0-58.0?C

Description

N-Boc-hydroxylamine is used in the preparation of azridines by cycloaddition of azides with nitroso Diels-Alder adducts. It acts as a reagent for the synthesis of hydroxylamine derivatives t-butyl-N-(acyloxy)carbamates and N,O-diacylated N-hydroxyarylsulfonamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-hydroxylamine is used in the preparation of azridines by cycloaddition of azides with nitroso Diels-Alder adducts. It acts as a reagent for the synthesis of hydroxylamine derivatives t-butyl-N-(acyloxy)carbamates and N,O-diacylated N-hydroxyarylsulfonamides.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases, reducing agents and alkali metals. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text