Thermo Scientific Chemicals

Lithium tetraborate trihydrate, 96%, Thermo Scientific Chemicals

Catalog number: A10358.22
100 g, Each
Thermo Scientific Chemicals

Lithium tetraborate trihydrate, 96%, Thermo Scientific Chemicals

Catalog number: A10358.22
100 g, Each
Quantity
Catalog number: A10358.22
also known as A10358-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37195-62-3
Appearance (Color)
White
Form
Powder or crystalline powder or granules
Assay (Non-aqueous acid-base Titration)
≥95.0 to ≤105.0%
Loss on Drying
21.03-27.16% (350°C/Constant weight) (2.5-3.5 waters)

Description

It is employed in the conductances of some potassium and sodium salts in dimethylformamide at 25°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the conductances of some potassium and sodium salts in dimethylformamide at 25°C.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text