2,4-Dichloro-6-methylpyrimidine is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Dichloro-6-methylpyrimidine is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine.
Solubility
Soluble in Chloroform, Ether, Ethyl Acetate and Toluene. Insoluble in water.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
- Eliud Hernández, et al. Novel inhibitors of Rac1 in metastatic breast cancer.P R Health Sci J.,2010,29(4), 348-56.
- Caroline Hadad, et al. 4-Arylvinyl-2,6-di(pyridin-2-yl)pyrimidines: synthesis and optical properties.J Org Chem.,2011,76(10), 3837-45.
- Wei Zhang. Fluorous synthesis of disubstituted pyrimidines.Org Lett.,2003,5(7), 1011-3.