Thermo Scientific Chemicals

2,4-Dichloro-6-methylpyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: A10363.22
100 g, Each
Thermo Scientific Chemicals

2,4-Dichloro-6-methylpyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: A10363.22
100 g, Each
Quantity
Catalog number: A10363.22
also known as A10363-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5424-21-5
Appearance (Color)
Pale yellow to yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
43.0-49.0?C

Description

2,4-Dichloro-6-methylpyrimidine is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichloro-6-methylpyrimidine is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine.

Solubility
Soluble in Chloroform, Ether, Ethyl Acetate and Toluene. Insoluble in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text