4-Fluorotoluene, 99%, Thermo Scientific Chemicals
4-Fluorotoluene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluorotoluene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A10365.22
also known as A10365-22
Price (USD)/ Each
39.80
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
39.80
Add to cart
4-Fluorotoluene, 99%, Thermo Scientific Chemicals
Catalog numberA10365.22
Price (USD)/ Each
39.80
-
Add to cart
Chemical Identifiers
CAS352-32-9
IUPAC Name1-fluoro-4-methylbenzene
Molecular FormulaC7H7F
InChI KeyWRWPPGUCZBJXKX-UHFFFAOYSA-N
SMILESCC1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.5%
Refractive Index1.4675-1.4705 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only