Thermo Scientific Chemicals

Triphenylmethanol, 98%, Thermo Scientific Chemicals

Catalog number: A10366.0B
1000 g, Each
Thermo Scientific Chemicals

Triphenylmethanol, 98%, Thermo Scientific Chemicals

Catalog number: A10366.0B
1000 g, Each
Quantity
Catalog number: A10366.0B
also known as A10366-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-84-6
IUPAC Name
triphenylmethanol
Molecular Formula
C19H16O
InChI Key
LZTRCELOJRDYMQ-UHFFFAOYSA-N
SMILES
OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Identification (FTIR)
Conforms
Melting Point (clear melt)
161-165?C
Solution Test
Clear to slightly hazy (5% ethanol)
Appearance (Color)
White to pale cream or pale yellow
Form
Crystalline powder

Description

Triphenylmethanol is used as a reagent in the research laboratory. It acts as an intermediate in the production of the commercially useful triarylmethane dyes. It is used in the preparation of triphenylmethane. It is also used as an antiproliferative agent. Further, it is used in the preparation of two-electron reduction product of pyrylogen. In addition to this, it reacts with triphenylphosphine oxide to form a 1:1 molecular complex. It serves as a specific clathrate host for methanol and dimethyl sulphoxide and forms clathrate inclusion complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylmethanol is used as a reagent in the research laboratory. It acts as an intermediate in the production of the commercially useful triarylmethane dyes. It is used in the preparation of triphenylmethane. It is also used as an antiproliferative agent. Further, it is used in the preparation of two-electron reduction product of pyrylogen. In addition to this, it reacts with triphenylphosphine oxide to form a 1:1 molecular complex. It serves as a specific clathrate host for methanol and dimethyl sulphoxide and forms clathrate inclusion complexes.

Solubility
Soluble in dioxane, ethanol, water, ether and benzene.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text