Thermo Scientific Chemicals

3-Phenylpropionaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: A10367.0B
1000 g, Each
Thermo Scientific Chemicals

3-Phenylpropionaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: A10367.0B
1000 g, Each
Quantity
Catalog number: A10367.0B
also known as A10367-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-53-0
IUPAC Name
3-phenylpropanal
Molecular Formula
C9H10O
InChI Key
YGCZTXZTJXYWCO-UHFFFAOYSA-N
SMILES
O=CCCC1=CC=CC=C1
Identification (FTIR)
Conforms
Refractive Index
1.5210-1.5300 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥94.0%

Description

3-Phenylpropionaldehyde is used as a flavoring agent. It is also used in the preparation of cinnamaldehyde, 2-chloro hydrocinnamaldehyde by alfa chlorination and mixture of homopropargyl alcohols. It serves as a precursor for the synthesis of mixture of syn- and anti-beta-hydroxyallylsilanes by hydroboration of allenylsilane. It is also involved in the catalytic asymmetric cyanosilylation to prepare (2S)-2-hydroxy-4-phenylbutanenitrile. Further, it undergoes Henry reaction to form nitroaldols by reacting with nitromethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenylpropionaldehyde is used as a flavoring agent. It is also used in the preparation of cinnamaldehyde, 2-chloro hydrocinnamaldehyde by alfa chlorination and mixture of homopropargyl alcohols. It serves as a precursor for the synthesis of mixture of syn- and anti-beta-hydroxyallylsilanes by hydroboration of allenylsilane. It is also involved in the catalytic asymmetric cyanosilylation to prepare (2S)-2-hydroxy-4-phenylbutanenitrile. Further, it undergoes Henry reaction to form nitroaldols by reacting with nitromethane.

Solubility
Miscible with chloroform, dichloromethane, ethyl acetate, alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text