Thermo Scientific Chemicals

4-Nitrophthalic acid, 97%, Thermo Scientific Chemicals

Catalog number: A10368.06
5 g, Each
Thermo Scientific Chemicals

4-Nitrophthalic acid, 97%, Thermo Scientific Chemicals

Catalog number: A10368.06
5 g, Each
Quantity
Catalog number: A10368.06
also known as A10368-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
610-27-5
IUPAC Name
4-nitrobenzene-1,2-dicarboxylic acid
Molecular Formula
C8H5NO6
InChI Key
SLBQXWXKPNIVSQ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(C=C1C(O)=O)[N+]([O-])=O
Appearance (Color)
Yellow or pale cream
Form
Powder or granules
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
158.5-164.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text