Thermo Scientific Chemicals

4-Nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A10369.30
250 g, Each
Thermo Scientific Chemicals

4-Nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A10369.30
250 g, Each
Quantity
Catalog number: A10369.30
also known as A10369-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-01-6
Melting Point (clear melt)
144-151°C
Form
Crystals or crystalline powder or powder or flakes
Identification (FTIR)
Conforms
Appearance (Color)
Yellow to dark brown
Assay (GC)
≥97.5%

Description

p-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.

Solubility
Soluble in hot water and alcohol

Notes
Store at 4°C. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text