Thermo Scientific Chemicals

2-Ethylhexylamine, 98%, Thermo Scientific Chemicals

Catalog number: A10372.AE
100 mL, Each
Thermo Scientific Chemicals

2-Ethylhexylamine, 98%, Thermo Scientific Chemicals

Catalog number: A10372.AE
100 mL, Each
Quantity
Catalog number: A10372.AE
also known as A10372-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-75-6
IUPAC Name
3-(aminomethyl)heptane
Molecular Formula
C8H19N
InChI Key
LTHNHFOGQMKPOV-UHFFFAOYNA-N
SMILES
CCCCC(CC)CN
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4290-1.4330 @ 20?C

Description

2-Ethylhexylamine is used to make pharmaceuticals and other chemicals. It is used as beam material and surface active agent. It also involves in synthesis of aliphatic amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexylamine is used to make pharmaceuticals and other chemicals. It is used as beam material and surface active agent. It also involves in synthesis of aliphatic amines.

Solubility
Soluble in water. 2.5 g/L (20°C)

Notes
Air Sensitive. Incompatible with strong oxidizing agents. Protect against electrostatic charges. Store away from acid chlorides and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text