Thermo Scientific Chemicals

1,2-Difluorobenzene, 98+%, Thermo Scientific Chemicals

Catalog number: A10377.18
50 g, Each
Thermo Scientific Chemicals

1,2-Difluorobenzene, 98+%, Thermo Scientific Chemicals

Catalog number: A10377.18
50 g, Each
Quantity
Catalog number: A10377.18
also known as A10377-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
367-11-3
IUPAC Name
1,2-difluorobenzene
Molecular Formula
C6H4F2
InChI Key
GOYDNIKZWGIXJT-UHFFFAOYSA-N
SMILES
FC1=CC=CC=C1F
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4420-1.4460 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid
Assay (GC)
≥98.0%

Description

1,2-Difluorobenzene(1,2-DFB) has been used to study the mechanism of dissociation of o-, m- and p-difluorobenzene ions by threshold photoelectron photoion coincidence spectroscopy. It has been used to study the room temperature adsorption of 1,2-DFB, 1,2-dichlorobenzene and 1,2-dibromobenzene on Si(100)2x1 by X-ray photoelectron spectroscopy and temperature programmed desorption. It is used as solvent in electrochemical studies on transition metal complexes. It is also applied as a solvent useful for electrochemical studies on transition metal complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Difluorobenzene(1,2-DFB) has been used to study the mechanism of dissociation of o-, m- and p-difluorobenzene ions by threshold photoelectron photoion coincidence spectroscopy. It has been used to study the room temperature adsorption of 1,2-DFB, 1,2-dichlorobenzene and 1,2-dibromobenzene on Si(100)2x1 by X-ray photoelectron spectroscopy and temperature programmed desorption. It is used as solvent in electrochemical studies on transition metal complexes. It is also applied as a solvent useful for electrochemical studies on transition metal complexes.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text