2-(Methylthio)benzoic acid is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Methylthio)benzoic acid is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.
Solubility
Soluble in Methanol.
Notes
Incompatible with strong oxidizing agents, acids, bases, amines. Store in a cool, dry place. Store in a tightly closed container.
RUO – Research Use Only
Yong Joon Kwon; Sang Bok Lee; Kwan Kim; Myung Soo Kim. Raman spectroscopy of 4-(methylthio)benzoic acid adsorbed on silver surfaces. Journal of molecular structure. 1994, 318 25-35 .