It is an interesting carbonyl equivalent and reacts with aldehydes or ketones to generate α-hydroxy carbonyl compounds. Benzothiazole is used as a chemical intermediate in organic synthesis. It is a precursor of rubber accelerators and a component of cyanine dyes. It is also used as a flavoring substance. It is also used as a flavor, antimicrobial agent, and component of cyanine dyes. Its derivatives are used as rubber accelerators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an interesting carbonyl equivalent and reacts with aldehydes or ketones to generate α-hydroxy carbonyl compounds. Benzothiazole is used as a chemical intermediate in organic synthesis. It is a precursor of rubber accelerators and a component of cyanine dyes. It is also used as a flavoring substance. It has been used as an antimicrobial agent in athletic shoe insoles. It is also used as a flavor, antimicrobial agent, and component of cyanine dyes. Its derivatives are used as rubber accelerators.
Solubility
Slightly soluble in water. Soluble in alcohol, carbon disulfide
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
- Nawrocki ST, et al. Comparative aquatic toxicity evaluation of 2-(thiocyanomethylthio)benzothiazole and selected degradation products using Ceriodaphnia dubia. Arch. Environ. Contam. Toxicol.,2005,48(3), 344-350.
- Asimakopoulos AG, et al. Occurrence and removal efficiencies of benzotriazoles and benzothiazoles in a wastewater treatment plant in Greece. The Science of the total environment.,2013,452-453163-171.
- n-BuLi gives the 2-lithio-derivative, stable below -50o, which behaves as a formyl anion equivalent, reacting with aldehydes and ketones. The derived alcohols have been transformed into various useful products, e.g. ɑ-hydroxy ketones: Tetrahedron Lett., 5, 9, 13, (1978); Bull. Chem. Soc. Jpn., 61, 3637 (1988):
- The lithio-derivative can also be acylated with esters, lactones, amides or nitriles. In these applications, benzothiazole is claimed to be superior to the better known 1,3-dithiane as a formyl anion synthon.
- See also 2-(Trimethyl silyl) benzothiazole, L11625, and 2-(Trimethyl silyl) thiazole, B21903.
- Reaction of the lithio-derivative at low temperatures with PCl3 gives, as the major product, 2,2'-bibenzothiazole instead of the expected tertiary phosphine: Heterocycles, 30, 347 (1990). POCl3 or SOCl2 also promote the symmetrical coupling reaction: Heteroatom. Chem., 5, 409 (1994).