Thermo Scientific Chemicals

Dodecanedioic acid, 99%, Thermo Scientific Chemicals

Catalog number: A10387.0B
1000 g, Each
Thermo Scientific Chemicals

Dodecanedioic acid, 99%, Thermo Scientific Chemicals

Catalog number: A10387.0B
1000 g, Each
Quantity
Catalog number: A10387.0B
also known as A10387-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
693-23-2
IUPAC Name
dodecanedioic acid
Molecular Formula
C12H22O4
InChI Key
TVIDDXQYHWJXFK-UHFFFAOYSA-N
SMILES
OC(=O)CCCCCCCCCCC(O)=O
Appearance (Color)
White to cream
Form
Crystalline powder or powder or flakes or pellets
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (Non-U.S. specification)
Assay (Silylated GC)
≥98.5% (Non-U.S. specification)
Assay from Suppliers CofA
≥98.5% (GC, U.S. specification)

Description

Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.

Solubility
Soluble in water, ethanol, hot methanol, hot toluene and hot acetic acid.

Notes
Incompatible with strong oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text