4-Ethylanisole, 98+%, Thermo Scientific Chemicals
4-Ethylanisole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Ethylanisole, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A10391.09
also known as A10391-09
Price (USD)/ Each
58.65
Online exclusive
65.00 
Save 6.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
58.65
Online exclusive
65.00 
Save 6.35 (10%)
Add to cart
4-Ethylanisole, 98+%, Thermo Scientific Chemicals
Catalog numberA10391.09
Price (USD)/ Each
58.65
Online exclusive
65.00 
Save 6.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1515-95-3
IUPAC Name1-ethyl-4-methoxybenzene
Molecular FormulaC9H12O
InChI KeyHDNRAPAFJLXKBV-UHFFFAOYSA-N
SMILESCCC1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5065-1.5105 @ 20°C
FormLiquid
Identification (FTIR)Conforms
It can be used the synthesis of 1-Methoxy-4-ethyl-1,4-cyclohexadiene. It is also applied in the ligand-assisted, copper-catalyzed enantioselective benzylic amination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used the synthesis of 1-Methoxy-4-ethyl-1,4-cyclohexadiene. It is also applied in the ligand-assisted, copper-catalyzed enantioselective benzylic amination.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Gaetano Cantarella, et al. Free radical versus electron-transfer routes of oxidation of hydrocarbons by laccase/mediator systems: Catalytic or stoichiometric procedures.J. Mol. Catal. B: Enzym.,2003,22(3-4), 135-144.
  2. Marten Klopstra, et al. Non-heme iron catalysts for the benzylic oxidation: a parallel ligand screening approach.Tetrahedron Lett.,2003,44(24), 4581-4584.