Thermo Scientific Chemicals

4-Ethylanisole, 98+%, Thermo Scientific Chemicals

Catalog number: A10391.18
50 g, Each
Thermo Scientific Chemicals

4-Ethylanisole, 98+%, Thermo Scientific Chemicals

Catalog number: A10391.18
50 g, Each
Quantity
Catalog number: A10391.18
also known as A10391-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1515-95-3
IUPAC Name
1-ethyl-4-methoxybenzene
Molecular Formula
C9H12O
InChI Key
HDNRAPAFJLXKBV-UHFFFAOYSA-N
SMILES
CCC1=CC=C(OC)C=C1
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.5065-1.5105 @ 20°C
Form
Liquid
Identification (FTIR)
Conforms

Description

It can be used the synthesis of 1-Methoxy-4-ethyl-1,4-cyclohexadiene. It is also applied in the ligand-assisted, copper-catalyzed enantioselective benzylic amination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used the synthesis of 1-Methoxy-4-ethyl-1,4-cyclohexadiene. It is also applied in the ligand-assisted, copper-catalyzed enantioselective benzylic amination.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text