Thermo Scientific Chemicals

2-Nitrobenzoic acid, 95%, Thermo Scientific Chemicals

Catalog number: A10393.36
500 g, Each
Thermo Scientific Chemicals

2-Nitrobenzoic acid, 95%, Thermo Scientific Chemicals

Catalog number: A10393.36
500 g, Each
Quantity
Catalog number: A10393.36
also known as A10393-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
552-16-9
IUPAC Name
2-nitrobenzoic acid
Molecular Formula
C7H5NO4
InChI Key
SLAMLWHELXOEJZ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1[N+]([O-])=O
Assay (Silylated GC)
≥94.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
143-149?C
Appearance (Color)
White to cream or pale yellow
Form
Crystals or powder or crystalline powder

Description

It is employed as a synthetic reagent and can function as a growth supplement for Pseudomonas fluorescens strain KU-7. It also finds its use as a reagent for protection of amine groups and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a synthetic reagent and can function as a growth supplement for Pseudomonas fluorescens strain KU-7. It also finds its use as a reagent for protection of amine groups and in organic synthesis.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text