Methyl 6-chloronicotinate, 98%, Thermo Scientific Chemicals
Methyl 6-chloronicotinate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 6-chloronicotinate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A10396.14
also known as A10396-14
Price (USD)/ Each
212.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
212.00
Add to cart
Methyl 6-chloronicotinate, 98%, Thermo Scientific Chemicals
Catalog numberA10396.14
Price (USD)/ Each
212.00
-
Add to cart
Chemical Identifiers
CAS73781-91-6
IUPAC Namemethyl 6-chloropyridine-3-carboxylate
Molecular FormulaC7H6ClNO2
InChI KeyRMEDXVIWDFLGES-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
FormCrystals or powder or crystalline powder
Melting Point (clear melt)82.0-89.0?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only