Thermo Scientific Chemicals

Dimethyl fumarate, 99%, Thermo Scientific Chemicals

Catalog number: A10402.30
250 g, Each
Thermo Scientific Chemicals

Dimethyl fumarate, 99%, Thermo Scientific Chemicals

Catalog number: A10402.30
250 g, Each
Quantity
Catalog number: A10402.30
also known as A10402-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
624-49-7
IUPAC Name
1,4-dimethyl (2E)-but-2-enedioate
Molecular Formula
C6H8O4
InChI Key
LDCRTTXIJACKKU-ONEGZZNKSA-N
SMILES
COC(=O)\C=C\C(=O)OC
Form
Crystals or crystalline powder or flakes
Free acid (titration)
≤1.5% as fumaric acid
Appearance (Color)
White to cream to pale brown
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%

Description

Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing

Solubility
Soluble in water (1.6 mg/ml at 20°C), methanol (30-36 mg/ml), ethanol (10 mg/ml at 25°C), DMSO (29 mg/ml at 25°C), and DMF (∼12 mg/ml).

Notes
Do not store together with acids. Incompatible with acids, bases, oxidizing agents, reducing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text