Thermo Scientific Chemicals

Methyl 2,5-dihydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A10407.06
5 g, Each
Thermo Scientific Chemicals

Methyl 2,5-dihydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A10407.06
5 g, Each
Quantity
Catalog number: A10407.06
also known as A10407-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2150-46-1
IUPAC Name
methyl 2,5-dihydroxybenzoate
Molecular Formula
C8H8O4
InChI Key
XGDPKUKRQHHZTH-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(O)=CC=C1O
Form
Powder
Melting Point (clear melt)
84.0-90.0?C
Assay (GC)
≥97.5%
Appearance (Color)
Pale cream to cream to pale brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text