Thermo Scientific Chemicals

1,4-Cyclohexanedione monoethylene acetal, 97%, Thermo Scientific Chemicals

Catalog number: A10408.06
5 g, Each
Thermo Scientific Chemicals

1,4-Cyclohexanedione monoethylene acetal, 97%, Thermo Scientific Chemicals

Catalog number: A10408.06
5 g, Each
Quantity
Catalog number: A10408.06
also known as A10408-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4746-97-8
IUPAC Name
1,4-dioxaspiro[4.5]decan-8-one
Molecular Formula
C8H12O3
InChI Key
VKRKCBWIVLSRBJ-UHFFFAOYSA-N
SMILES
O=C1CCC2(CC1)OCCO2
Appearance (Color)
White to pale brown
Form
crystals or crystalline powder
Solution Test
10% w/v in acetone : Clear
Assay (GC)
> 96.0%

Description

1,4-Cyclohexanedione is used in the preparation of series of potent analgesic compounds. 1,4-Cyclohexanedione is also used as a building block in the synthesis of tritium labelled probes for the autoradiography study of the dopamine reuptake complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Cyclohexanedione is used in the preparation of series of potent analgesic compounds. 1,4-Cyclohexanedione is also used as a building block in the synthesis of tritium labelled probes for the autoradiography study of the dopamine reuptake complex.

Solubility
Soluble in chloroform & methanol.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents and strong acids. Ensure adequate ventilation during use.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text