Thermo Scientific Chemicals

Ninhydrin, 99%, Thermo Scientific Chemicals

Catalog number: A10409.14
25 g, Each
Thermo Scientific Chemicals

Ninhydrin, 99%, Thermo Scientific Chemicals

Catalog number: A10409.14
25 g, Each
Quantity
Catalog number: A10409.14
also known as A10409-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
485-47-2
IUPAC Name
2,2-dihydroxy-2,3-dihydro-1H-indene-1,3-dione
Molecular Formula
C9H6O4
InChI Key
FEMOMIGRRWSMCU-UHFFFAOYSA-N
SMILES
OC1(O)C(=O)C2=CC=CC=C2C1=O
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
7.5-11.6% (0.8-1.3 waters)
Appearance (Color)
White to cream or pale yellow
Identification (FTIR)
Conforms
Assay (HPLC)
≥98.5%

Description

Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.

Solubility
Soluble in water, acetone, ethanol, and methanol. Slightly soluble in chloroform and ether.

Notes
Light Sensitive. Incompatible with strong oxidizing agents, amines, alkalis and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text