Thermo Scientific Chemicals

Lithium sulfate monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: A10410.0E
2500 g, Each
Thermo Scientific Chemicals

Lithium sulfate monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: A10410.0E
2500 g, Each
Quantity
Catalog number: A10410.0E
also known as A10410-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10102-25-7
IUPAC Name
dilithium(1+) hydrate sulfate
Molecular Formula
H2Li2O5S
InChI Key
RKGLUDFWIKNKMX-UHFFFAOYSA-L
SMILES
[Li+].[Li+].O.[O-]S([O-])(=O)=O
Appearance (Color)
White
Water Content (Karl Fischer Titration)
12.9-16.4% (0.9-1.2 waters of hydration)
Assay (unspecified)
≥98.5 to ≤101.5% (Dry wt. basis)
Form
Crystalline powder

Description

Lithium sulfate is used to treat bipolar disorder. It is also used in ultrasound-type non-destructive testing because they are very efficient sound generators. It is also used in building blocks and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium sulfate is used to treat bipolar disorder. It is also used in ultrasound-type non-destructive testing because they are very efficient sound generators. It is also used in building blocks and chemical synthesis.

Solubility
Soluble in water. 340 g/L (20 C)

Notes
Store under dry inert gas. Protect from humidity and water. Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text