Lithium sulfate monohydrate, 99%, Thermo Scientific Chemicals
Lithium sulfate monohydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium sulfate monohydrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number A10410.36
also known as A10410-36
Price (USD)/ Each
94.65
Online exclusive
105.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
94.65
Online exclusive
105.00 
Save 10.35 (10%)
Add to cart
Lithium sulfate monohydrate, 99%, Thermo Scientific Chemicals
Catalog numberA10410.36
Price (USD)/ Each
94.65
Online exclusive
105.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10102-25-7
IUPAC Namedilithium(1+) hydrate sulfate
Molecular FormulaH2Li2O5S
InChI KeyRKGLUDFWIKNKMX-UHFFFAOYSA-L
SMILES[Li+].[Li+].O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Water Content (Karl Fischer Titration)12.9-16.4% (0.9-1.2 waters of hydration)
Assay (unspecified)≥98.5 to ≤101.5% (Dry wt. basis)
FormCrystalline powder
Lithium sulfate is used to treat bipolar disorder. It is also used in ultrasound-type non-destructive testing because they are very efficient sound generators. It is also used in building blocks and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium sulfate is used to treat bipolar disorder. It is also used in ultrasound-type non-destructive testing because they are very efficient sound generators. It is also used in building blocks and chemical synthesis.

Solubility
Soluble in water. 340 g/L (20 C)

Notes
Store under dry inert gas. Protect from humidity and water. Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only
H. G. Smith; S. W. Peterson and H. A. Levy. Neutron-Diffraction Study of Lithium Sulfate Monohydrate. J. Chem. Phys. 1968, 48 5561.