Thermo Scientific Chemicals

L-Histidine, 98+%

Catalog number: A10413.22
100 g, Each
Thermo Scientific Chemicals

L-Histidine, 98+%

Catalog number: A10413.22
100 g, Each
Quantity
Catalog number: A10413.22
also known as A10413-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71-00-1
IUPAC Name
2-amino-3-(1H-imidazol-5-yl)propanoic acid
Molecular Formula
C6H9N3O2
InChI Key
HNDVDQJCIGZPNO-UHFFFAOYNA-N
SMILES
NC(CC1=CN=CN1)C(O)=O
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Optical Rotation
-39 ? 2? (c = 5 in water)
Identification (FTIR)
Conforms
Appearance (Color)
White

Description

L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.

Solubility
Soluble in water (15 mg/ml at 20°C).

Notes
Store at 4°C.Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text