2,2,3,3,4,4,4-Heptafluoro-1-butanol, 98%, Thermo Scientific Chemicals
2,2,3,3,4,4,4-Heptafluoro-1-butanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,3,3,4,4,4-Heptafluoro-1-butanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
50 g
250 g
Catalog number A10414.14
also known as A10414-14
Price (USD)/ Each
133.65
Online exclusive
148.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
133.65
Online exclusive
148.00 
Save 14.35 (10%)
Add to cart
2,2,3,3,4,4,4-Heptafluoro-1-butanol, 98%, Thermo Scientific Chemicals
Catalog numberA10414.14
Price (USD)/ Each
133.65
Online exclusive
148.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS375-01-9
IUPAC Name2,2,3,3,4,4,4-heptafluorobutan-1-ol
Molecular FormulaC4H3F7O
InChI KeyWXJFKAZDSQLPBX-UHFFFAOYSA-N
SMILESOCC(F)(F)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Assay (GC)≥97.5%
FormLiquid
It is employed as a halogenated solvent in electrospray liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a halogenated solvent in electrospray liquid chromatography.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
RUO – Research Use Only

General References:

  1. Yolanda Díaz-de-Mera, et al.Atmospheric chemistry of HFE-7000 (CF3CF2CF2OCH3) and 2,2,3,3,4,4,4-heptafluoro-1-butanol (CF3CF2CF2CH2OH): kinetic rate coefficients and temperature dependence of reactions with chlorine atoms.Environmental Science and Pollution Research.,2008,15(7), 584-591.
  2. Richard B. van Breemen. Electrospray Liquid Chromatography-Mass Spectrometry of Carotenoids.Anal. Chem.,1995,67(13), 2004-2009.