Thermo Scientific Chemicals

2-Fluoro-m-xylene, 98%, Thermo Scientific Chemicals

Catalog number: A10424.14
25 g, Each
Thermo Scientific Chemicals

2-Fluoro-m-xylene, 98%, Thermo Scientific Chemicals

Catalog number: A10424.14
25 g, Each
Quantity
Catalog number: A10424.14
also known as A10424-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
443-88-9
IUPAC Name
2-fluoro-1,3-dimethylbenzene
Molecular Formula
C8H9F
InChI Key
JTAUTNBVFDTYTI-UHFFFAOYSA-N
SMILES
CC1=CC=CC(C)=C1F
Assay (GC)
≥97.5%
Form
Liquid
Appearance (Color)
Clear colorless
Refractive Index
1.4780-1.4820 @ 20?C

Description

It is used in the spectroscopic observation of jet-cooled 2-fluoro-m-xylyl radical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the spectroscopic observation of jet-cooled 2-fluoro-m-xylyl radical.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text