Thermo Scientific Chemicals

Azobenzene, 97+%, Thermo Scientific Chemicals

Catalog number: A10425.36
500 g, Each
Thermo Scientific Chemicals

Azobenzene, 97+%, Thermo Scientific Chemicals

Catalog number: A10425.36
500 g, Each
Quantity
Catalog number: A10425.36
also known as A10425-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-33-3
Appearance (Color)
Orange to red
Assay (GC)
≥97.0%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or chunks
Melting Point (clear melt)
62.0-72.0?C

Description

Azobenzene is used to construct photoswitch tethered ligands to turn protein activity on and off. It is also used to increase the extracellular or intracellular concentration of active molecules to study the mechanism of action of sympathetic and parasympathetic neuromediators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azobenzene is used to construct photoswitch tethered ligands to turn protein activity on and off. It is also used to increase the extracellular or intracellular concentration of active molecules to study the mechanism of action of sympathetic and parasympathetic neuromediators.

Solubility
Soluble in alcohol, ether, benzene and glacial acetic acid. Insoluble in water.

Notes
Store at 4°C. Incompatible with strong oxidizing agents. Air and light sensitive.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text